Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCNN1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577817
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577817 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577817 Supplier Invitrogen™ Supplier No. PA577817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

1 μg/mL of PA5-77817 was sufficient for detection of beta-ENaC in 20 μg of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 87kDa.

Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception. Heterotetramer of two alpha, one beta and one gamma subunit. A delta subunit can replace the alpha subunit. Interacts with the WW domains of NEDD4, NEDD4L, WWP1 and WWP2. Defects in SCNN1B are a cause of autosomal recessive pseudohypoaldosteronism type 1 (PHA1) [MIM:264350]. PHA1 is a rare salt wasting disease resulting from target organ unresponsiveness to mineralocorticoids. There are 2 forms of PHA1: the autosomal recessive form that is severe, and the dominant form which is more milder and due to defects in mineralocorticoid receptor. Autosomal recessive PHA1 is characterized by an often fulminant presentation in the neonatal period with dehydration, hyponatraemia, hyperkalaemia, metabolic acidosis, failure to thrive and weight loss. Defects in SCNN1B are a cause of Liddle syndrome. It is an autosomal dominant disorder characterized by pseudoaldosteronism and hypertension associated with hypokalemic alkalosis. The disease is caused by constitutive activation of the renal epithelial sodium channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCNN1B
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene SCNN1B
Gene Accession No. P37090, P51168, Q9WU38
Gene Alias Amiloride-sensitive sodium channel subunit beta; amiloride-sensitive sodium channel subunit beta 1; amiloride-sensitive sodium channel subunit beta-like protein; BESC1; Beta ENaC; beta-ENaC; Beta-NaCH; betaxENaC; ENaC beta; ENaCb; enacbeta; epithelial Na(+) channel subunit beta; epithelial sodium channel beta-2 subunit; epithelial sodium channel beta-3 subunit; epithelial sodium channel, nonvoltage-gated 1, beta; nasal epithelial sodium channel beta subunit; nonvoltage-gated sodium channel 1 subunit beta; RNENACB; SCNEB; SCNN 1B; SCNN1B; scnn1b.L; scnn1b-a; scnn1b-b; sodium channel epithelial 1 beta subunit; sodium channel, non voltage gated 1 beta subunit; sodium channel, non voltage gated 1 beta subunit L homeolog; sodium channel, nonvoltage-gated 1 beta; sodium channel, nonvoltage-gated 1, beta; sodium channel, non-voltage-gated 1, beta subunit; sodium channel, nonvoltage-gated, type I, beta; XELAEV_18045378mg
Gene Symbols SCNN1B, scnn1b.L
Host Species Rabbit
Immunogen Produced against the C-terminal tail (amino acids 617-638) of rat beta ENaC (antibody designation 3755-2).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100754564, 20277, 24767, 378536, 6338
Target Species Human, Mouse, Rat, Hamster, Frog
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.