Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCNN1D Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142418
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000. Preliminary analysis showed no signal in Western Blot.
SNX6 interacts with members of the transforming growth factor-beta family of receptor serine-threonine kinases. These receptors belong to two classes: type II receptors that bind ligand, and type I receptors that are subsequently recruited to transduce the signal. Of the type II receptors, SNX6 was found to interact strongly with ActRIIB and more moderately with wild type and kinase-defective mutants of TbetaRII. Of the type I receptors, SNX6 was found to interact only with inactivated TbetaRI. SNXs 1-4 also interacted with the transforming growth factor-beta receptor family, showing different receptor preferences. Conversely, SNX6 behaved similarly to the other SNX proteins in its interactions with receptor tyrosine kinases. Strong heteromeric interactions were also seen among SNX1, -2, -4, and -6, suggesting the formation in vivo of oligomeric complexes. These findings are the first evidence for the association of the SNX family of molecules with receptor serine-threonine kinases.
Specifications
| SCNN1D | |
| Polyclonal | |
| Unconjugated | |
| SCNN1D | |
| Amiloride-sensitive sodium channel subunit delta; delta-ENaC; Delta-NaCH; DNACH; ENaCd; ENaCdelta; epithelial Na(+) channel subunit delta; MGC149710; Nonvoltage-gated sodium channel 1 subunit delta; SCNED; SCNN1D; sodium channel epithelial 1 delta subunit; sodium channel, non voltage gated 1 delta subunit; sodium channel, nonvoltage-gated 1, delta; sodium channel, non-voltage-gated 1, delta subunit; sodium channel, voltage-gated, type I, delta polypeptide | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6339 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P51172 | |
| SCNN1D | |
| Peptide with sequence C-AGPQPLETLDT. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction