Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCNN1D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530864
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SNX6 interacts with members of the transforming growth factor-beta family of receptor serine-threonine kinases. These receptors belong to two classes: type II receptors that bind ligand, and type I receptors that are subsequently recruited to transduce the signal. Of the type II receptors, SNX6 was found to interact strongly with ActRIIB and more moderately with wild type and kinase-defective mutants of TbetaRII. Of the type I receptors, SNX6 was found to interact only with inactivated TbetaRI. SNXs 1-4 also interacted with the transforming growth factor-beta receptor family, showing different receptor preferences. Conversely, SNX6 behaved similarly to the other SNX proteins in its interactions with receptor tyrosine kinases. Strong heteromeric interactions were also seen among SNX1, -2, -4, and -6, suggesting the formation in vivo of oligomeric complexes. These findings are the first evidence for the association of the SNX family of molecules with receptor serine-threonine kinases.
Specifications
SCNN1D | |
Polyclonal | |
Unconjugated | |
SCNN1D | |
Amiloride-sensitive sodium channel subunit delta; delta-ENaC; Delta-NaCH; DNACH; ENaCd; ENaCdelta; epithelial Na(+) channel subunit delta; MGC149710; Nonvoltage-gated sodium channel 1 subunit delta; SCNED; SCNN1D; sodium channel epithelial 1 delta subunit; sodium channel, non voltage gated 1 delta subunit; sodium channel, nonvoltage-gated 1, delta; sodium channel, non-voltage-gated 1, delta subunit; sodium channel, voltage-gated, type I, delta polypeptide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6339 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P51172 | |
SCNN1D | |
Recombinant fragment corresponding to a region within amino acids 109 and 442 of Human SCNN1D. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction