Learn More
Invitrogen™ SCO1 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20345 can be used with blocking peptide PEP-0465.
Specifications
Specifications
| Antigen | SCO1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SCO1 |
| Gene Accession No. | O75880, Q5SUC9 |
| Gene Alias | 2610001C07Rik; D11Bwg1310e; Protein SCO1 homolog, mitochondrial; RGD1559538; SCO cytochrome c oxidase assembly protein 1; SCO cytochrome oxidase deficient homolog 1; SCO cytochrome oxidase deficient homolog 1 (yeast); SCO1; SCO1 cytochrome c oxidase assembly protein; SCO1, cytochrome c oxidase assembly protein; SCOD1; synthesis of cytochrome C oxidase 1 |
| Gene Symbols | SCO1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.