Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCOC Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP172058
Description
SCOC Polyclonal specifically detects SCOC in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
SCOC | |
Polyclonal | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_001146956.1 | |
SCOC | |
Peptide with sequence C-HTLEDLSARVDAVKE corresponding to internal region (near C-Terminus) according to NP_001147135.1, NP_001147024.1, NP_115936.2, NP_001147107.1, NP_001147162.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize all six reported isoforms (NP_001146956.1; NP_001147135.1; NP_001147024.1; NP_115936.2; NP_001147107.1; NP_001147162.1). Reported variants represent identical protein: NP_001147057.1, NP_001146918.1, NP_115936.2 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
HRIHFB2072, SCOCOshort coiled coil protein, short coiled-coil protein | |
Goat | |
Affinity Purified | |
RUO | |
60592 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction