Learn More
Beckman Coulter Scopio X100 Series Full-Field Digital Cell Morphology Scanner
Catalog No. p-200106332 Shop All Beckman Coulter Products
Click to view available options
Product Line:
Scopio X100
Scopio X100HT
The digital cell morphology platforms featuring Full-Field technology are transforming cell morphology analysis.
Full-Field Digital Imaging
  • Dynamic Scan Image providing enhanced assessment quality and added context for comprehensive analysis.
  • A seamless integration of CBC results before and during slide review, alongside Scopio's differential
  • No trade-off between field-of-view and resolution, allowing for assessment and analysis of cell morphology at an unmatched scale and depth
AI powered Decision Support
  • Adaptive Monolayer Detection algorithm, designed to identify the clinically relevant area and adapt for optimal review.
  • White Blood Cells DSS for detection and pre-classification into 14 classes.
  • Detection of platelets and pre-estimation from 10 fields of view, including the identification of platelet clumps throughout the scan area and the feathered edge.
Seamless LIS Integration
  • Advanced digital platform promotes remote review and consultation throughout the secure hospital network.
  • Streamline workflows with an enhanced review process and minimized logistical constraints
  • Help specialists collaborate across the secured hospital network with easy remote reviews and consultations
  • Customize the analysis process for department-specific needs with features like multiple slide review, customizable category naming, seamless LIS integration, and flexible reporting options
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Beckman Coulter Analyzers
Product Line Scopio X100
Throughput 15 Slides/hr.
Includes Processing Unit and Full-Field PBS
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.