Learn More
Eisco™ Scoria Specimen
SCI_ED
Catalog No. S24770 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24770 1 Ea. 2 to 3 cm
S26614 10/Pk. 2 to 3 cm
S24771 12/Pk. 2 to 3 cm
S26650 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24770 Supplier Eisco™ Supplier No. ESNG0050
Add to Cart
Add to Cart
Scoria forms when spatter from bubbly mafic lava cools quickly.
  • For use in lessons on eruption style and cooling rate
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Erupted from spatter cone as large air bubbles made their way through magma to surface
  • Crystallization happens so quickly that large air pockets are trapped within rock, often giving samples jagged, sharp appearance
Specifications
Product Type Igneous Rock
For Use With (Application) Lessons on eruption style and cooling rate
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Scoria