Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Scoria Specimen
SCI_ED

Catalog No. S26650 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S26650 1 Ea. 7 to 8 cm
S24770 1 Ea. 2 to 3 cm
S26614 10/Pk. 2 to 3 cm
S24771 12/Pk. 2 to 3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26650 Supplier Eisco™ Supplier No. ESNG0050HAND
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Scoria forms when spatter from bubbly mafic lava cools quickly.

  • For use in lessons on eruption style and cooling rate
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Erupted from spatter cone as large air bubbles made their way through magma to surface
  • Crystallization happens so quickly that large air pockets are trapped within rock, often giving samples jagged, sharp appearance

Specifications

Product Type Igneous Rock
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Specimen Type Scoria
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.