Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Glass Cloth Electrical Tape

Catalog No. p-7182403
Click to view available options
Dimensions (LxW):
66 ft. x 1/2 in.
66 ft. x 3/4 in.

Scotch™ Glass Cloth Electrical Tape is premium grade and edge tear resistant.

  • Printable
  • Non corrosive adhesive
  • Offers solvent resistant protection
  • Flame retardant and suitable for 600V rated, dry location application motors and transformers.
  • Thermosetting, silicon adhesive provides an increased bond at high temperatures
  • Glass cloth backing optimally resists resist shrinking and burning
  • Used as a coil cover, anchor, banding, core, layer and crossover insulation
  • Printable and provides high mechanical strength and excellent performance by withstanding high temperatures
  • Withstands a temperature range of 32 to 392°F (0 to 200°C)
  • UL Listed, Mil-I-19166C, CSA Certified and RoHS 2011/65/EU Compliant
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Silicone
Product Type Glass Cloth Tape
For Use With (Application) Automotive, Commericial Solutions, Communications, Consumer, Design and Construction, Electronics, Energy, Health Care, Manufacturing, Mining, Oil and Gas
Model 69-1/2″X66′
Dimensions (LxW) 66 ft. x 1/2 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.