Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Vinyl Electrical Tape Super 88

Catalog No. 19072079 Shop All 3M Products
Click to view available options
Dimensions (LxW):
36 yd. x 3/4 in.
66 ft. x 3/4 in.
Quantity:
10 Rolls/Carton, 100 Rolls/Cs.
12 Rolls/pk., 48 Rolls/Cs.

3M™ Scotch™ Vinyl Electrical Tape Super 88 features a combination of pressure-sensitive, rubber-resin adhesive and PVC backing that offers excellent electrical and mechanical protection

  • 8.5 mil thick
  • Premium grade
  • All-weather
  • Provides high resistance to UV rays, abrasion, moisture, alkalis, acids, corrosion and varying weather conditions
  • Provides a protective jacket and electrical insulation for cables and splices up to 600V
  • Made of durable PVC
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance UL Listed, CSA Certified and RoHS 2011/65/EU Compliant
Color Black
Material Rubber
Product Type Vinyl Electrical Tape
For Use With (Application) Electrical Maintenance
Model 88-3/4X66FT
Quantity 10 Rolls/Carton, 100 Rolls/Cs.
Temperature Range (English) 0°F to 221°F
Temperature Range (Metric) 18°C to 105°C
Dimensions (LxW) 66 ft. x 3/4 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.