Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott's Bluing Reagent (Scott's Tapwater Substitute), Bluing Agent for Nuclear Hematoxylin Stain, Ricca Chemical
SDP

Catalog No. p-7239371 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Bluing Agent for Nuclear Hematoxylin Stain

Specifications

Physical Form Liquid
Chemical Name or Material Scott's Bluing Reagent
Name Note Scott's Tapwater Substitute
CAS 7732-18-5, 10034-99-8, 144-55-8, 50-00-0, 67-56-1
CAS Min % 95.0, 1.91, 0.51, 0.19
CAS Max % 99.0, 1.99, 0.53, 0.2
Packaging Natural Poly Bottle
Color Colorless
Quantity 1 L
Grade Laboratory
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.