Learn More
Scott Safety™ Air-Pak Fifty™ SCBA Respirator Thrust Washers
Catalog No. p-4910172
Change view
Click to view available options
Thickness:
0.005 in.
0.030 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness
19-818-492 0.005 in.
19-818-491 0.030 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-818-492 Supplier Scott Safety™ Supplier No. 7012473137
Only null left
Add to Cart
Add to Cart
Thrust washer replacement parts are thin, disk-shaped components located between a rotating surface and a stationary component to help keep shaft assemblies aligned.
  • Thrust washers fit between bearings and stationary surfaces along a shaft
  • Support axial and thrust movement
  • Keep shaft assemblies aligned and stop uneven load distribution
  • Available in two thicknesses
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Air-Pak fifty SCBA
Material Metal
Model 3502-03
Product Type Thrust Washer
Thickness 0.005 in.