Learn More
Scott Safety™ AV-3000™ Facepiece
Description
Requires
To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister
- Top-Down Convertibility allows single facepiece for APR, PAPR, SAR and SCBA respirators
- One fit test and reduced training
- All with Comfort Seal with deep chin cup for reliable, comfortable fit
- Mechanical and electronic communication options across all applications
- All components latex free
- Four-point Kevlar™, polyester or rubber head harness options
Order Info
Fit testing required prior to using this facepiece. Prescription lens kit (805753-01) and other accessories available separately
Certifications
All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.
Compliance
All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007.
Specifications
Specifications
| Certifications/Compliance | NIOSH, ANSI Z87.1-2003, NFPA 1981:2007 |
| Material | Dupont™ Kevlar™ aramid fiber, Polyisoprene Seal |
| Product Type | Facepiece |
| Format | Large |
| For Use With (Equipment) | Av-3000™ facepiece |
| Harness Style | Dupont Kevlar aramid fibers |
| Includes | Large dual voicemitters |
| Latex Free | Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.