Learn More
Scott Safety™ SKA-PAK™ Supplied Air Respirators
Catalog No. 19122013
Click to view available options
Includes:
AV-2000 (CS) Poly facepiece
AV-2000, Dupont™ Kevlar™ aramid fiber (CS) facepiece
No Facepiece
This item is not returnable. View return policy
This item is not returnable. View return policy
Offers emergency respiratory protection for variety of applications.
  • Designed for use in hazardous atmospheres
  • Combination Type C positive pressure respirator with escape cylinder for entry into/escape from hazardous, confined space or IDLH situations
  • Includes E-Z Flo pressure demand regulator with first-breath activation and low exhalation
  • 5-, 10-, or 15-minute rated cylinder egress options for selection versatility based on task at hand
  • Hip-mounted cylinder with durable, secure cylinder band retention system
  • Available in two harness styles:
  • Polypropylene - chemically resistant and lightweight
  • Kevlar - Forhigh heat environments or when there is potential for sparking
  • Available with Hansen, Schrader, Cejn, Hansen HK plug or Hansen HK socket fittings
  • Can be used with AV-2000, AV-3000 or AV-3000 with SureSeal facepieces
Applications:
Maintenance work, hazardous materials handling, turn-arounds and confined space operations.Used in chemical, food processing, petrochemical, oil & gas, ship building and public utilities.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Supplied Air Respirator System
For Use With (Application) Maintenance work, hazardous materials handling, turn-arounds and confi ned space operations, chemical, food processing, petrochemical, oil & gas, ship building and public utilities
Includes AV-2000, Dupont™ Kevlar™ aramid fiber (CS) facepiece
Pressure 2216 psig
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.