Learn More
Scott Safety™ Speed-Evac™ Mouthbit Respirator
Catalog No. p-59707
Change view
Click to view available options
For Use With (Application):
Acid gas, non IDLH environments
Includes:
Disposable plastic pouch
Quantity:
1 Ea.
Catalog No. For Use With (Application) Includes Quantity
19-818-562 Acid gas, non IDLH environments Disposable plastic pouch 1 Ea.
1 options
Catalog No. 19-818-562 Supplier Scott Safety™ Supplier No. 80383401
Designed for emergency escape respiratory protection against a variety of toxic gases in low level concentrations
  • Small, lightweight and ergonmically designed so it can be donned for immediate emergency escape protection
  • One size fits all
  • Requires minimal training and is completely disposable
  • comfortable mouthbit, chin rest ane easy-to-use nose clip
  • Choice of disposable plastic pouch or reusable ABS case
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH
Product Type Mouth-Bit Respirator
For Use With (Application) Acid gas, non IDLH environments
Includes Disposable plastic pouch
Quantity 1 Ea.
Size Universal