Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek
SDP

Catalog No. 06802414 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Concentration:
10 ppm
100 ppm
1000 ppm
15 ppm
99.80%
CO 0.5%, CO2 0.5%, H2 0.5%, O2 0.5% In nitrogen
CO 1%, CO2 1%, H2 1%, O2 1% in Nitrogen
CO 1%, CO2 1%, Methane 1%, Ethane 1%, Ethylene 1%, Ecetylene 1% in Nitrogen
Methane 1% in nitrogen
THC less than 1ppm
Components:
C1-C6 n-Paraffins in Helium
C1-C6 n-Paraffins in Nitrogen
C2-C6 Olefins in Nitrogen
Carbon Dioxide
Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen
Carbon Monoxide, Carbon Dioxide, Methane, Ethane, Ethylene, Ecetylene in Nitrogen
Methane in Nitrogen
Methane, Ethane, Ethylene, Acetylene, Propane, Propylene, n-Butane, Propyne in Nitrogen
Trichloroethylene in Nitrogen
Vinyl Chloride in Nitrogen
Zero Air
n-Butane, Isobutane, cis-2-Butene, trans-2-Butene, 1-Butene, Isobutene, 1,3-Butadiene, Ethyl Acetylene in Nitrogen
Type:
Air Standard
Transportable Air Standard
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Components Type
06-802-414 THC less than 1ppm Zero Air Air Standard
06-802-417 99.80% Carbon Dioxide Air Standard
06-802-448 Methane 1% in nitrogen Methane in Nitrogen Air Standard
06-802-464 10 ppm Vinyl Chloride in Nitrogen Air Standard
06-802-470 CO 0.5%, CO2 0.5%, H2 0.5%, O2 0.5% In nitrogen Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen Transportable Air Standard
06-802-476 CO 1%, CO2 1%, Methane 1%, Ethane 1%, Ethylene 1%, Ecetylene 1% in Nitrogen Carbon Monoxide, Carbon Dioxide, Methane, Ethane, Ethylene, Ecetylene in Nitrogen Transportable Air Standard
06-802-473 CO 1%, CO2 1%, H2 1%, O2 1% in Nitrogen Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen Transportable Air Standard
06-802-490 1000 ppm C1-C6 n-Paraffins in Helium Transportable Air Standard
06-802-493 100 ppm C1-C6 n-Paraffins in Nitrogen Transportable Air Standard
06-802-497 100 ppm C2-C6 Olefins in Nitrogen Transportable Air Standard
06-802-504 15 ppm n-Butane, Isobutane, cis-2-Butene, trans-2-Butene, 1-Butene, Isobutene, 1,3-Butadiene, Ethyl Acetylene in Nitrogen Transportable Air Standard
06-802-502 15 ppm Methane, Ethane, Ethylene, Acetylene, Propane, Propylene, n-Butane, Propyne in Nitrogen Transportable Air Standard
06-802-460 10 ppm Trichloroethylene in Nitrogen Air Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 06-802-414 Supplier Restek Corporation Supplier No. 34449
Add to Cart
Add to Cart

Ideal for peak identification work

  • CGA 165 connection and can deliver large volumes of sample
  • Scotty™ (Air Liquide) 48
  • Contents: 48L
  • Pressure: 300 psig (21 bar)
  • Outlet Fitting: CGA 165
  • Weight: 1.75 lb. (0.8kg)
  • Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
  • DOT Specifications: 39 NRC

All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements

Specifications

Certifications/Compliance DOT 39 NRC
Components Zero Air
Diameter (Metric) 10.2 cm
Diameter (English) 4 in.
Concentration THC less than 1ppm
Volume (Metric) Gas 48 L
For Use With (Application) Ideal for peak identification work
Height (English) 16.25 in.
Height (Metric) 41 cm
Pressure 300 psi
Weight (English) 1.75 lb.
Volume (English) Gas 12.68 gal.
Weight (Metric) 0.8 kg
Type Air Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.