Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InBios International SCoV-2 Ag Detect™ Rapid Test
SDP

Catalog No. 22004203
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-004-203 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-004-203 Supplier InBios International Supplier No. COVAF50
Add to Cart
Add to Cart
This item is not returnable. View return policy

The SCoV-2 Ag Detect™ Rapid Test is a lateral flow chromatographic immunoassay intended for the qualitative detection of the SARS-CoV-2 nucleocapsid protein antigen in direct anterior nasal swab specimens from individuals who are suspected of COVID-19 by their healthcare provider.

  • Point of Care: CLIA Waived test can be performed in a variety of settings, from physician offices to school health clinics, with symptomatic individuals. Do not use for individuals who have no symptoms or have had symptoms longer than 4 days
  • Sequence analysis performed at InBios suggests test will detect variants currently in circulation, including KP.3.1.1
  • Identifies acute infection in symptomatic patients: PPA (Relative Sensitivity): 84.2%, NPA (Relative Specificity): 99.8%
  • Unique patent pending design – no mixing step
  • Quick visual results – no instrumentation needed
  • 50 test pack, includes positive and negative controls
  • Room temperature storage
  • 100% manufactured in the USA

Specifications

Certifications/Compliance FDA 510(k) Cleared
Content And Storage 20 to 30°C
CPT Code 87811
CE Marker CE-IVD
Clia Complexity Waived
Detectable Analytes SARS-CoV-2
Detection Method Visual
DoA Calibrators No
Type rapid test
Disposable Single-use
Includes 50 tests, 50 swabs, 4 vials buffer, postive control swab
Quantity 50 Tests
Sterility Non-sterile
Form Swab
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.