Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCP1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300229B
Description
SCP1 Polyclonal specifically detects SCP1 in Mouse, Rat, Chicken, Mammal, Parasite, Monkey, Human (Negative) samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
SCP1 | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry 1:750, Immunocytochemistry/Immunofluorescence 1:100-1:750, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:750 | |
Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Mouse, Rat, Chicken, Mammalia, Parasite, Monkey, Human (Negative) | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SYCP1 | |
A synthetic peptide made to the C-terminus of the mouse SCP1 protein sequence. [UniProt# Q62209] | |
0.1 mL | |
Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
6847 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction