Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146254 Supplier Invitrogen™ Supplier No. PA146254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The target sequence has 100% sequence homology with rat.

Major component of the transverse filaments of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCP1
Applications ChIP Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7-8
Gene SYCP1
Gene Accession No. Q03410, Q15431, Q62209
Gene Alias A coiled-coil related protein specific for synapsed regions of meiotic prophase chromosomes; AW742678; cancer/testis antigen 8; CT8; DlkI; FA1; HOM-TES-14; Ly107; MGC104417; Peg9; pG2; pref-1; SCP1; SCP-1; Sycp1; Synaptonemal complex protein 1; testicular tissue protein Li 191; ZOG
Gene Symbols SYCP1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of the mouse SCP1 protein sequence.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20957, 25276, 6847, 772081
Target Species Human, Mouse, Rat, Monkey, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.