Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCP2 Monoclonal Antibody (OTI1D2)
GREENER_CHOICE

Catalog No. PIMA526336
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526336 Supplier Invitrogen™ Supplier No. MA526336
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCP2 Monoclonal Antibody for IHC (P), Western Blot

This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCP2
Gene Accession No. P22307
Gene Alias AA409774; AA409893; C76618; C79031; NLTP; nonspecific lipid transfer protein; Non-specific lipid-transfer protein; NSLIPTR; ns-LTP; NSL-TP; propanoyl-CoA C-acyltransferase; SCP2; SCP-2; SCP-CHI; SCPX; SCP-X; Sterol carrier protein 2; sterol carrier protein 2, liver; sterol carrier protein X
Gene Symbols SCP2
Host Species Mouse
Immunogen Full length human recombinant protein of SCP2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6342
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.