Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCP3 Mouse anti-Human, Clone: 1A11F1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SCP3 |
---|---|
Clone | 1A11F1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6836
![]() ![]() |
Proteintech Group Inc
664091IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregcy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
SCP3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COR1, SCP 3, SCP3, SYCP3, synaptonemal complex protein 3 | |
SYCP3 | |
IgG1 | |
Protein G | |
Antibody |
1A11F1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q8IZU3 | |
50511 | |
SCP3 Fusion Protein Ag19330 | |
Primary | |
-20°C | |
SYCP3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title