Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA131226
Description
PA1-31226 detects SCP3 from human, mouse samples.
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregnancy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
SCP3 | |
Polyclonal | |
Unconjugated | |
Sycp3 | |
Cor1; LOC615896 protein; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; Sycp3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100523378, 101099735, 20962, 25561, 418097, 50511, 615896 | |
4°C, do not freeze | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P70281, Q3T0E2, Q63520, Q8IZU3 | |
Sycp3 | |
Synthetic peptide (Human) (C terminal). | |
100 μL | |
Primary | |
Human, Mouse, Rat, Bovine, Pig, Feline, Chicken | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction