Learn More
Scp3 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Scp3 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2346
![]() ![]() |
Rockland Immunochemicals
600401MP9 |
100 μg |
Each for $636.24
|
|
|||||
50-199-2347
![]() ![]() |
Rockland Immunochemicals
600401MP9S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse Scp3. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse and golden hamster and 93% to rat.
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregnancy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
Scp3 | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
P70281 | |
20962 | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
BOS_5289; Cor1; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; SYCP3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like | |
Sycp3 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Sycp3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.