Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCP3/SYCP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 31 publication
$243.50 - $499.50
Specifications
Antigen | SCP3/SYCP3 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot, Immunohistochemistry 1:750, Immunocytochemistry/Immunofluorescence 1:100 - 1:750, Immunoprecipitation reported in scientific literature (PMID 27932493), Immunohistochemistry-Paraffin 1:750, Immunohistochemistry-Frozen |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300231SS
![]() |
Novus Biologicals
NB300231SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB300231
![]() |
Novus Biologicals
NB300231 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
SCP3/SYCP3 | |
Western Blot, Immunohistochemistry 1:750, Immunocytochemistry/Immunofluorescence 1:100 - 1:750, Immunoprecipitation reported in scientific literature (PMID 27932493), Immunohistochemistry-Paraffin 1:750, Immunohistochemistry-Frozen | |
Polyclonal | |
Rabbit | |
Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
50511 | |
A synthetic peptide made to the C-terminal region of the mouse SCP3 protein. [UniProt# P70281] | |
Primary | |
Store at 4C. Do not freeze. |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Chicken | |
MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 | |
SYCP3 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title