Learn More
SCP3/SYCP3 Antibody - BSA Free, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 56 publications
Supplier:  Novus Biologicals NB300232
 View more versions of this product
Catalog No. NB300232
Only null left
Add to Cart
Description
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Feline, Parasite samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
Specifications
Specifications
SCP3/SYCP3
Polyclonal
Unconjugated
PBS with 0.09% Sodium Azide
anti-SCP3, MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3
Rabbit
Affinity Purified
RUO
Primary
Human, rat, pig, bovine and mouse. Other species have not been tested.
Store at 4C. Do not freeze.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 0.5 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation reported in scientific literature (PMID 18694876), Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:200-1:500, Immunoblotting reported in scientific literature (PMID 27612028)
Q8IZU3
SYCP3
A synthetic peptide made to the C-terminal region of the human SCP3 protein. [UniProt# Q8IZU3]
0.1 mL
Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
50511
Mouse, Rat, Pig, Bovine, Chicken, Feline, Parasite
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only