Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCP3/SYCP3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 56 publications
Supplier: Novus Biologicals NB300232SS
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Feline, Parasite samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
SCP3/SYCP3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
anti-SCP3, MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, rat, pig, bovine and mouse. Other species have not been tested. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 0.5 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation reported in scientific literature (PMID 18694876), Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:200-1:500, Immunoblotting reported in scientific literature (PMID 27612028) | |
Q8IZU3 | |
SYCP3 | |
A synthetic peptide made to the C-terminal region of the human SCP3 protein. [UniProt# Q8IZU3] | |
0.025 mL | |
Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
50511 | |
Mouse, Rat, Pig, Bovine, Chicken, Feline, Parasite | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction