Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCP3/SYCP3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP278133V
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SCP3/SYCP3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SYCP3 | |
Partial synthetic peptide made to an N-terminal region of the human SCP3 protein (between amino acids 10-80) [UniProt Q8IZU3] | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
Human, Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction