Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCP3/SYCP3, Rabbit anti-Human,Mouse, DyLight 755, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP278133IR
Description
SCP3/SYCP3 Polyclonal antibody specifically detects SCP3/SYCP3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
SCP3/SYCP3 | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Store at 4°C in the dark. | |
Polyclonal |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Affinity Purified, Conjugated | |
SYCP3 | |
Partial synthetic peptide made to an N-terminal region of the human SCP3 protein (between amino acids 10-80) [UniProt Q8IZU3] | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
Primary | |
Human, Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title