Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Screw Cap for Externally Threaded Sample Tubes (T501 and T502)

Catalog No. 23045268 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Natural
Orange
Red
White
Includes:
Lip seal
Silicone washer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
23-045-268 Natural Lip seal
23-045-269 Orange Silicone washer
23-045-270 Red Silicone washer
23-045-271 White Silicone washer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-045-268 Supplier Simport™ Scientific Supplier No. T501N
Add to Cart
Add to Cart
This item is not returnable. View return policy

Screw Cap for Externally Threaded Sample Tubes (T501 and T502) features long skirt and super-fast thread design that allows it to be removed or sealed with single turn.

  • Made of polypropylene
  • Autoclavable
  • For externally threaded tubes
  • Tubes sold separately

Specifications

Includes Lip seal
Product Type Screw Cap
Material Polypropylene
Color Natural
No. per Pack 1000 Pk.
Sterility Non-sterile
For Use With (Equipment) T501 and T502 Series Sample Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.