Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Screw Caps for Internally Threaded Sample Tubes (T500)

Catalog No. 23045267 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-267 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-045-267 Supplier Simport™ Scientific Supplier No. T500N
Add to Cart
Add to Cart

Screw Caps for Internally Threaded Sample Tubes (T500) are available in wide variety of colors.

  • Made of polypropylene
  • Autoclavable
  • For internally threaded tubes
  • Tubes sold separately

Specifications

Pre-sterilized No
Product Type Screw Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Sterility Non-sterile
For Use With (Equipment) Internally Threaded Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.