Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Screw Vial Caps and Seals

Catalog No. 03250640
Click to view available options
Color:
Black
White, Blue
Septum:
Red PTFE/white silicone seal

Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.

Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.

TRUSTED_SUSTAINABILITY

Specifications

Description Blue cap with red PTFE/white silicone seal; Thermo Scientific, PE, Claris, Shimadzu, Varian
Color White, Blue
Material PTFE, Silicone
Septum Red PTFE/white silicone seal
For Use With (Equipment) Thermo Scientific TRACE/AS3000/AI3000/AS800/TriPlus GC, PE Autosystem/AS-2000, Claris 500, ISS-100
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.