Learn More
Globe Scientific Screwcap Sample Tubes, Non-Graduated
Catalog No. 21402915
Change view
Click to view available options
Bottom Shape:
Conical (Self-standing)
Round
Round (Self-standing)
Capacity (Metric):
1.2 mL
2 mL
3 mL
4 mL
5 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape Capacity (Metric)
21-402-915 Conical (Self-standing) 1.2 mL
21-402-916 Round 2 mL
21-402-917 Round (Self-standing) 2 mL
21-402-918 Round (Self-standing) 3 mL
21-402-919 Round 4 mL
21-402-920 Round (Self-standing) 4 mL
21-402-921 Round (Self-standing) 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 21-402-915 Supplier Globe Scientific Supplier No. 6030
Only null left
Add to Cart
Add to Cart
These tubes and caps have been designed for the storage and transportation of biological material, with non graduated
  • Tubes are manufactured from non-toxic polypropylene and offer strength and clarity with non-graduated
  • The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
  • The caps are available with or with out a silicone o-ring and are available in a variety of colors
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Specifications
Autoclavable Autoclavable
Capacity (Metric) 1.2 mL
Bottom Shape Conical (Self-standing)
Material Polypropylene
Thread Style External
Sterility Non-sterile
Graduated No
Product Type Sample Tube