Learn More
Globe Scientific Screwcap Sample Tubes, Non-Graduated
Description
- Tubes are manufactured from non-toxic polypropylene and offer strength and clarity with non-graduated
- The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
- The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
- This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
- The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
- The caps are available with or with out a silicone o-ring and are available in a variety of colors
- The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
- This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
- The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 4 mL |
| Bottom Shape | Round |
| Material | Polypropylene |
| Thread Style | External |
| Sterility | Non-sterile |
| Graduated | No |
| Product Type | Sample Tube |