Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRIB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566680 Supplier Invitrogen™ Supplier No. PA566680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLLEVNQQSLL GLTHGEAVQL LRSVGDTLTV LVCDGFEAST DAALEVSPGV IANPFAAGIG HRNSLESISS IDRE Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRIB
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCRIB
Gene Accession No. Q14160
Gene Alias AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul
Gene Symbols SCRIB
Host Species Rabbit
Immunogen Recombinant Human SCRIB. Recombinant protein control fragment (Product #RP-107328).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23513
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.