Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRIB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142476
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142476 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142476 Supplier Invitrogen™ Supplier No. PA5142476
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_874365.3; NP_056171.3). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.

SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRIB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SCRIB
Gene Accession No. Q14160
Gene Alias AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul
Gene Symbols SCRIB
Host Species Goat
Immunogen Peptide with sequence C-PEGPGKEKELPGQ.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23513
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.