Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCRIB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554821
Description
Immunogen sequence: VSVIQFLEAP IGDEDAEEAA AEKRGLQRRA TPHPSELKVM KRSIEGRRSE ACPCQPDSGS PLPAEEEKRL SAESGLSEDS RPSASTVS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
Specifications
SCRIB | |
Polyclonal | |
Unconjugated | |
SCRIB | |
AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23513 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.40 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14160 | |
SCRIB | |
Recombinant protein corresponding to Human SCRIB. Recombinant protein control fragment (Product #RP-93933). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction