Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRN1 Monoclonal Antibody (OTI1E4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526337

Catalog No. PIMA526337


Only null left
Add to Cart

Description

Description

SCRN1 Monoclonal Antibody for Western Blot, IHC (P)

This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCRN1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q12765, Q6AY84
SCRN1
Full length human recombinant protein of SCRN1 produced in HEK293T cell.
100 μL
Primary
Human, Rat, Monkey
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1E4
Unconjugated
SCRN1
2810019K23Rik; 6330535A03Rik; AI852905; Kiaa0193; mKIAA0193; Scrn1; secernin 1; secernin-1; SES1
Mouse
Affinity Chromatography
RUO
502776, 9805
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.