Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582886
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582886 Supplier Invitrogen™ Supplier No. PA582886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKLVPKTQSP CFGDDDPAKK EPRFQEKPDR RHELYKAHEW ARAIIESDQE QGRKLRSTML ELEKQGLEAM EEILTSSEPL DPAEVGDLFY DCVD Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 90%.

This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCRN1
Gene Accession No. Q12765
Gene Alias 2810019K23Rik; 6330535A03Rik; AI852905; Kiaa0193; mKIAA0193; Scrn1; secernin 1; secernin-1; SES1
Gene Symbols SCRN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCRN1. Recombinant protein control fragment (Product #RP-92726).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.