Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCRN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | SCRN1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119405
|
Invitrogen™
PA5119405 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121853
|
Invitrogen™
PA5121853 |
20 μL |
Each for $209.50
|
|
|||||
Description
SCRN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.Specifications
SCRN1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
SCRN1 | |
2810019K23Rik; 6330535A03Rik; AI852905; Kiaa0193; mKIAA0193; Scrn1; secernin 1; secernin-1; SES1 | |
SCRN1 | |
Primary | |
Antigen Affinity Chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q12765 | |
9805 | |
Recombinant protein Secernin 1. The antigen corresponds to amino acid range 2-414 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title