Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588321 Supplier Invitrogen™ Supplier No. PA588321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRLWAAQRIQ EGARNISNQL SIGTDISAQH PELRTHAQAK GWWDGQGAFD FAQIFSLTQQ PVRMEAAKAR FQAGRELLRQ RQGGITAEVM MGILRDKESG ICMDSGGFRT TASMVSVLPQ DPTQPCVHFL TATPDPSRSV FKPFIFGMGV AQAPQVLSPT FGAQDPVRTL PRFQTQVDRR HTLYRGHQAA LGLMERDQDR GQQLQQKQQD LEQEGLEATQ GLLAGEWAPP LWELGSLFQA FVKRESQAYA; Positive Samples: BxPC-3, HeLa, A-375, Mouse liver, Mouse kidney, Mouse intestine.

SCRN2 is a member of the secernin family of proteins which includes the homologous SCRN1, a protein that was first identified as being involved in the regulation of exocytosis from peritoneal mast cells. Little is known of SCRN2, but studies have shown that SCRN1 may possess epitopes that function as tumor-associated antigens in gastric cancers and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN2 may also be involved in protein secretion or the regulation of cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SCRN2
Gene Accession No. Q8VCA8, Q96FV2
Gene Alias AV001119; D11Moh48; SCRN2; secernin 2; secernin-2; Ses2
Gene Symbols SCRN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 176-425 of human SCRN2 (NP_6123642).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217140, 90507
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.