Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCRN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557016
Description
Immunogen sequence: RRHPLYQKHQ QALEVVNNNE EKAKIMLDNM RKLEKELFRE MESILQNKHL DVEKIVNLFP QCTKDEIQIY QSNLSVKVS Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 65%.
SCRN3 is a member of the secernin family of proteins which includes the homologous SCRN1, a protein that was first identified as being involved in the regulation of exocytosis from peritoneal mast cells. Little is known of SCRN3, but studies have shown that SCRN1 may possess epitopes that function as tumor-associated antigens in gastric cancers and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN3 may also be involved in protein secretion or the regulation of cell growth.
Specifications
SCRN3 | |
Polyclonal | |
Unconjugated | |
Scrn3 | |
4833415E20Rik; AI427019; AV002033; Scrn3; secernin 3; secernin-3; Ses3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0VDG4 | |
Scrn3 | |
Recombinant protein corresponding to Human SCRN3. Recombinant protein control fragment (Product #RP-102629). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction