Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557017 Supplier Invitrogen™ Supplier No. PA557017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DMRNYAKRKG WWDGKKEFDF AAAYSYLDTA KMMTSSGRYC EGYKLLNKHK GNITFETMME ILRDKPSGIN MEGEFLTTA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

SCRN3 is a member of the secernin family of proteins which includes the homologous SCRN1, a protein that was first identified as being involved in the regulation of exocytosis from peritoneal mast cells. Little is known of SCRN3, but studies have shown that SCRN1 may possess epitopes that function as tumor-associated antigens in gastric cancers and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN3 may also be involved in protein secretion or the regulation of cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRN3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Scrn3
Gene Accession No. Q0VDG4
Gene Alias 4833415E20Rik; AI427019; AV002033; Scrn3; secernin 3; secernin-3; Ses3
Gene Symbols Scrn3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCRN3. Recombinant protein control fragment (Product #RP-96126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79634
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.