Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589813 Supplier Invitrogen™ Supplier No. PA589813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVCDEEALLG MDLVRLGLER ADTAEKALNV IVDLLEKYGQ GGNCTEGRMV FSYHNSFLIA DRNEAWILET AGKYWAAEKV QEGVRNISNQ LSITTKIARE HPDMRNYAKR KGWWDGKKEF DFAAAYSYLD TAKMMTSSGR YCEGYKLLNK HKGNITFETM MEILRDKPSG INMEGEFLTT ASMVSILPQD SSLPCIHFFT GTPDPERSVF KPFIFVPHIS QLLDTSSPTF ELEDLVKKKS HFKPDRRHPL YQKHQQALEV VNNNEEKAKI MLDNMRKLEK ELFREMESIL QNKHLDVEKI VNLFPQCTKD E; Positive Samples: HepG2, A-549, 293T.

SCRN3 is a member of the secernin family of proteins which includes the homologous SCRN1, a protein that was first identified as being involved in the regulation of exocytosis from peritoneal mast cells. Little is known of SCRN3, but studies have shown that SCRN1 may possess epitopes that function as tumor-associated antigens in gastric cancers and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN3 may also be involved in protein secretion or the regulation of cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRN3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Scrn3
Gene Accession No. Q0VDG4, Q3TMH2
Gene Alias 4833415E20Rik; AI427019; AV002033; Scrn3; secernin 3; secernin-3; Ses3
Gene Symbols Scrn3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-410 of human SCRN3 (NP_0788592).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311731, 74616, 79634
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.