Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCRO Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987310
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7310 Supplier Bioss Supplier No. BS6523R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Part of an E3 ubiquitin ligase complex for neddylation. Required for neddylation of cullin components of E3 cullin-RING ubiquitin ligase complexes by enhancing the rate of cullins neddylation. Functions to recruit the NEDD8-charged E2 enzyme to the cullin component. Involved in the release of inhibitory effets of CAND1 on cullin-RING ligase E3 complex assembly and activity. Acts also as an oncogene facilitating malignant transformation and carcinogenic progression.

Specifications

Antigen SCRO
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DCUN1D1
Gene Accession No. Q96GG9
Gene Alias DCN1; DCN1, defective in cullin neddylation 1, domain containing 1; DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae); DCN1-like protein 1; DCNL1; DCUN1 domain-containing protein 1; Dcun1d1; DCUN1L1; defective in cullin neddylation 1 domain containing 1; Defective in cullin neddylation protein 1-like protein 1; pTes3; Rp42; RP42 homolog; SCCRO; SCRO; squamous cell carcinoma related oncogene; Squamous cell carcinoma-related oncogene; Tes3; testis derived transcript 3; Testis-specific protein 3
Gene Symbols DCUN1D1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SCRO/DCUN1D1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54165
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.