Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCRT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566946 Supplier Invitrogen™ Supplier No. PA566946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APEEYSDPESP QSSLSARYFR GEAAVTDSYS MDAF Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRT2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCRT2
Gene Accession No. Q9NQ03
Gene Alias AW060659; FP7030; ortholog of human scratch homolog 2, zinc finger protein SCRT2; RGD1564796; scratch 2 protein; scratch family transcriptional repressor 2; scratch family zinc finger 2; scratch homolog 2 zinc finger protein; scratch homolog 2, zinc finger protein; scratch homolog 2, zinc finger protein (Drosophila); Scrt2; Transcriptional repressor scratch 2; ZNF898B
Gene Symbols SCRT2
Host Species Rabbit
Immunogen Recombinant Human SCRT2. Recombinant protein control fragment (Product #RP-107596).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85508
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.