Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCTR Monoclonal Antibody (3H1)
GREENER_CHOICE

Catalog No. PIMA519902
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519902 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519902 Supplier Invitrogen™ Supplier No. MA519902
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: CDVLQVLWEE QDQCLQELSR EQTGDLGTEQ PVPGCEGMWD NISCWPSSVP GRMVEVECPR FLRMLTSRNG SLFRNCTQDG WSETFPRPNL ACGVNVNDSS NEKRHSYLLK.

The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCTR
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3H1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Sctr
Gene Accession No. P47872
Gene Alias 6530402O03Rik; MYOP; pancreatic secretin receptor; Sctr; SCT-R; Secretin receptor; SR
Gene Symbols Sctr
Host Species Mouse
Immunogen SCTR (NP_002971, 32 a.a. ~ 141 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6344
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.