Learn More
Invitrogen™ SCUBE1 Polyclonal Antibody

Description
A suggested positive control is Daudi cell lysate. PA5-20990 can be used with blocking peptide PEP-1104.
Specifications
Specifications
| Antigen | SCUBE1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SCUBE1 |
| Gene Accession No. | Q8IWY4 |
| Gene Alias | 7330410C13Rik; A630023E24Rik; im:7427863; SCUBE1; signal peptide; Signal peptide, CUB and EGF-like domain-containing protein 1; signal peptide, CUB and EGF-like domain-containing protein 1; LOW QUALITY PROTEIN: signal peptide, CUB and EGF-like domain-containing protein 1; signal peptide, CUB domain and EGF like domain containing 1; signal peptide, CUB domain, EGF-like 1; signal peptide-CUB domain-EGF-related 1 |
| Gene Symbols | SCUBE1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.