Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCUBE2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169603
Description
SCUBE2 Polyclonal specifically detects SCUBE2 in Human samples. It is validated for Western Blot.Specifications
| SCUBE2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Cegb1, Cegf1, CEGP1, FLJ16792, FLJ35234, MGC133057, Protein CEGP1, signal peptide, CUB and EGF-like domain-containing protein 2, signal peptide, CUB domain, EGF-like 2 | |
| Rabbit | |
| 110 kDa | |
| 100 μL | |
| Primary | |
| Mouse: 86%; Zebrafish: 83%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NQ36 | |
| SCUBE2 | |
| Synthetic peptides corresponding to SCUBE2(signal peptide, CUB domain, EGF-like 2) The peptide sequence was selected from the C terminal of SCUBE2. Peptide sequence KTPEAWNMSECGGLCQPGEYSADGFAPCQLCALGTFQPEAGRTSCFPCGG. | |
| Affinity purified | |
| RUO | |
| 57758 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction