Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCUBE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556278 Supplier Invitrogen™ Supplier No. PA556278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGIHLSSDVT TIRTSVTFKL NEGKCSLKNA ELFPEGLRPA LPEKHSSVKE SFRYVNLTCS SGKQVPGAPG RPSTPKEMFI TVEFELETNQ KEVTASCDLS CIVKRTEKRL RKAIRTLRKA VHREQFHLQL SGMNLDVAK Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.

SCUBE2 is a member of a family of secreted glycoproteins that contain N-terminal EGF-like repeats and C-terminal cysteine-rich motifs and CUB domain and are expressed in embryonic neuroectoderm and vascular epithelium. SCUBE2 is a close homolog to SCUBE1 and can form a heterodimeric complex with SCUBE1. SCUBE2 is thought to participate in the Hedgehog signaling pathway by specifically interacting with Sonic Hedgehog (SHH) and the receptor PTCH1, enhancing the SHH signaling activity in the raft microdomains of the plasma membrane, and may thus play important roles in organ development and tumor progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCUBE2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCUBE2
Gene Accession No. Q9NQ36
Gene Alias CEGB1; CEGF1; CEGP1; Protein CEGP1; RGD1563998; Scube/You; SCUBE2; Scube2-ps1; signal peptide; Signal peptide, CUB and EGF-like domain-containing protein 2; signal peptide, CUB domain and EGF like domain containing 2; signal peptide, CUB domain, EGF-like 2; signal peptide, CUB domain, EGF-like 2, pseudogene 1
Gene Symbols SCUBE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCUBE2. Recombinant protein control fragment (Product #RP-88796).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57758
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.