Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCUBE3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2411220.025MG
Description
SCUBE3 Polyclonal specifically detects SCUBE3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SCUBE3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
SCUBE3 | |
Antibody was raised against a 15 amino acid peptide near the center of human SCUBE3. The immunogen is located within amino acids 440 - 490 of SCUBE3. Amino Acid Squence: AAPARAGHNGNSTNS | |
Affinity Purified | |
RUO | |
222663 | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
CEGF3, CUB and EGF containing protein 3, CUB domain and EGF-like repeat containing 3, DKFZp686B09105, DKFZp686B1223, DKFZp686D20108, FLJ34743, signal peptide, CUB and EGF-like domain containing protein 3, signal peptide, CUB and EGF-like domain-containing protein 3, signal peptide, CUB domain, EGF-like 3 | |
Rabbit | |
109 kDa | |
0.025 mg | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction